Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131029.2 | NP_571104.1 | 3 - 4 | 1863 | 81 | |
GenBank mRNA | AF002018.1 | - | 3 - 4 | 636 | 81 | |
AF002717.1 | - | 3 - 4 | 1735 | 81 | ||
CU638812.1 | - | 3 - 4 | 1735 | 81 | ||
KM655584.1 | - | 3 - 4 | 1356 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |