Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131252.1 | NP_571327.1 | 7 - 8 | 637 | 82 | |
XM_005158170.3 | XP_005158227.1 | 7 - 8 | 685 | 82 | ||
GenBank mRNA | BC044403.1 | - | 7 - 8 | 637 | 82 | |
BC067595.1 | - | 6 - 7 | 554 | 82 | ||
BC165881.1 | - | 4 - 5 | 395 | 82 | ||
S76875.1 | - | 3 - 4 | 177 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |