Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: spon1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131515.1 | NP_571590.1 | 15 - 16 | 2415 | 67 | |
GenBank mRNA | AB006086.1 | - | 15 - 16 | 2415 | 67 | |
BC162586.1 | - | 15 - 16 | 2398 | 67 | ||
KM655720.1 | - | 15 - 16 | 2263 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |