Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tradd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131607.2 | NP_571682.1 | 5 - 6 | 701 | 94 | |
GenBank mRNA | BC083425.1 | - | 5 - 6 | 701 | 94 | |
BC164053.1 | - | 4 - 5 | 620 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |