Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: u2af2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029956.1 | NP_001025127.1 | 7 - 8 | 1211 | 66 | |
XM_005157895.4 | XP_005157952.1 | 7 - 8 | 1285 | 66 | ||
XM_021466696.1 | XP_021322371.1 | 7 - 8 | 1307 | 66 | ||
GenBank mRNA | BC098548.1 | - | 7 - 8 | 1211 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |