Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stxbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089376.1 | NP_001082845.1 | 12 - 13 | 1067 | 110 | |
GenBank mRNA | BC171526.1 | - | 12 - 13 | 1049 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |