Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc4a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198338.1 | NP_938152.1 | 15 - 16 | 1930 | 72 | |
GenBank mRNA | AF350072.1 | - | 15 - 16 | 1930 | 72 | |
AY098936.1 | - | 15 - 16 | 72 | |||
BC093126.1 | - | 8 - 9 | 72 | |||
BC134843.1 | - | 2 - 3 | 72 | |||
BC163932.1 | - | 15 - 16 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |