Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndrg3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199797.1 | NP_956091.1 | 2 - 3 | 176 | 86 | |
XM_005161986.2 | XP_005162043.1 | 2 - 3 | 270 | 86 | ||
XM_009296689.1 | XP_009294964.1 | 2 - 3 | 270 | 86 | ||
XM_009296691.2 | XP_009294966.1 | 2 - 3 | 270 | 86 | ||
XM_017353522.1 | XP_017209011.1 | 2 - 3 | 270 | 86 | ||
GenBank mRNA | BC044139.1 | - | 2 - 3 | 176 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |