Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnppd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199928.1 | NP_956222.1 | 7 - 8 | 776 | 119 | |
GenBank mRNA | BC044514.1 | - | 7 - 8 | 776 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |