Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: thap12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213081.1 | NP_998246.1 | 2 - 3 | 272 | 106 | |
GenBank mRNA | BC044456.1 | - | 2 - 3 | 272 | 106 | |
BC165660.1 | - | 2 - 3 | 249 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |