Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsm14ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214772.2 | NP_999937.2 | 8 - 9 | 1247 | 97 | |
XM_005158973.3 | XP_005159030.1 | 9 - 10 | 1346 | 97 | ||
XM_005158974.3 | XP_005159031.1 | 9 - 10 | 1346 | 97 | ||
GenBank mRNA | AY577000.1 | - | 8 - 9 | 1224 | 97 | |
BC047185.1 | - | 8 - 9 | 1244 | 97 | ||
BC065685.1 | - | 8 - 9 | 1212 | 97 | ||
BC165583.1 | - | 8 - 9 | 1106 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |