Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3bb.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025450.1 | NP_001020621.1 | 3 - 4 | 312 | 149 | |
XM_005162028.4 | XP_005162085.1 | 3 - 4 | 516 | 149 | ||
XM_009296716.3 | XP_009294991.1 | 3 - 4 | 514 | 149 | ||
XM_009296717.3 | XP_009294992.1 | 3 - 4 | 520 | 149 | ||
GenBank mRNA | AB196915.1 | - | 3 - 4 | 312 | 149 | |
BC124098.1 | - | 3 - 4 | 396 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |