Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044933.2 | NP_001038398.2 | 26 - 27 | 3119 | 86 | |
XM_005160726.4 | XP_005160783.1 | 26 - 27 | 3141 | 86 | ||
GenBank mRNA | BC129209.1 | - | 26 - 27 | 3119 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |