Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099245.1 | NP_001092715.1 | 3 - 4 | 450 | 122 | |
GenBank mRNA | BC146745.1 | - | 3 - 4 | 450 | 122 | |
HM210101.1 | - | 3 - 4 | 416 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |