Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131555.2 | NP_571630.2 | 7 - 8 | 1478 | 106 | |
GenBank mRNA | AF280090.1 | - | 7 - 8 | 1417 | 106 | |
BC129260.1 | - | 7 - 8 | 1478 | 106 | ||
BC164222.1 | - | 7 - 8 | 1399 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |