Search Thermo Fisher Scientific
Gene Symbol: sema3ga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014823.1 | NP_001014823.1 | 6 - 7 | 677 | 81 | |
XM_005167031.4 | XP_005167088.1 | 7 - 8 | 1312 | 81 | ||
GenBank mRNA | AY766120.1 | - | 6 - 7 | 677 | 81 | |
BC163710.1 | - | 6 - 7 | 761 | 81 | ||
BC163715.1 | - | 6 - 7 | 761 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |