Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hadhaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105276.1 | NP_001098746.1 | 17 - 18 | 1898 | 69 | |
GenBank mRNA | BC093195.1 | - | 17 - 18 | 1888 | 69 | |
BC115212.1 | - | 17 - 18 | 1898 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |