Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amy2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213011.2 | NP_998176.2 | 8 - 9 | 1252 | 70 | |
GenBank mRNA | BC054669.1 | - | 8 - 9 | 1227 | 70 | |
BC062867.1 | - | 8 - 9 | 1229 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |