Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sirt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002071.1 | NP_001002071.1 | 6 - 7 | 707 | 66 | |
XM_021479673.1 | XP_021335348.1 | 6 - 7 | 724 | 66 | ||
XM_021479674.1 | XP_021335349.1 | 6 - 7 | 569 | 66 | ||
XM_021479677.1 | XP_021335352.1 | 4 - 5 | 449 | 66 | ||
GenBank mRNA | BC071405.1 | - | 6 - 7 | 707 | 66 | |
BC153591.1 | - | 6 - 7 | 674 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |