Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dmbx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017625.2 | NP_001017625.1 | 3 - 4 | 508 | 106 | |
GenBank mRNA | BC093284.1 | - | 3 - 4 | 450 | 106 | |
BC164462.1 | - | 2 - 3 | 364 | 106 | ||
DQ317638.1 | - | 3 - 4 | 508 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |