Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rxrab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131153.1 | NP_571228.1 | 4 - 5 | 596 | 88 | |
XM_009301955.3 | XP_009300230.1 | 6 - 7 | 1077 | 88 | ||
GenBank mRNA | U29894.1 | - | 4 - 5 | 596 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |