Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3s10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199820.2 | NP_956114.2 | 3 - 4 | 469 | 85 | |
XM_005156611.2 | XP_005156668.1 | 3 - 4 | 380 | 85 | ||
GenBank mRNA | BC059196.1 | - | 3 - 4 | 481 | 85 | |
BC066670.1 | - | 3 - 4 | 469 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |