Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbp1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212895.2 | NP_998060.2 | 2 - 3 | 340 | 83 | |
GenBank mRNA | AY395732.1 | - | 2 - 3 | 343 | 83 | |
BC153598.1 | - | 2 - 3 | 390 | 83 | ||
BC162113.1 | - | 2 - 3 | 323 | 83 | ||
BC162130.1 | - | 2 - 3 | 323 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |