Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007065.1 | NP_001007066.1 | 21 - 22 | 2403 | 97 | |
XM_017358267.2 | XP_017213756.1 | 20 - 21 | 2398 | 97 | ||
GenBank mRNA | AY677196.1 | - | 21 - 22 | 2403 | 97 | |
EU826643.1 | - | 20 - 21 | 2372 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |