Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020680.2 | NP_001018516.1 | 3 - 4 | 243 | 76 | |
GenBank mRNA | BC095759.1 | - | 3 - 4 | 245 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |