Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cables1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105665.1 | NP_001099135.1 | 9 - 10 | 1504 | 77 | |
XM_005163651.4 | XP_005163708.1 | 8 - 9 | 1951 | 77 | ||
XM_021478424.1 | XP_021334099.1 | 8 - 9 | 1954 | 77 | ||
GenBank mRNA | EF105292.1 | - | 9 - 10 | 1504 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |