Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: smfn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200744.2 | NP_957038.2 | 3 - 4 | 478 | 62 | |
GenBank mRNA | BC059540.1 | - | 3 - 4 | 362 | 62 | |
EH454655.1 | - | 3 - 4 | 385 | 62 | ||
EH469615.1 | - | 4 - 5 | 756 | 62 | ||
EH580645.1 | - | 3 - 4 | 512 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |