Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacna1da | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203484.1 | NP_982351.1 | 47 - 48 | 6162 | 110 | |
XM_021479576.1 | XP_021335251.1 | 48 - 49 | 6383 | 110 | ||
XM_021479577.1 | XP_021335252.1 | 48 - 49 | 6383 | 110 | ||
XM_021479578.1 | XP_021335253.1 | 48 - 49 | 6380 | 110 | ||
XM_021479579.1 | XP_021335254.1 | 47 - 48 | 6356 | 110 | ||
XM_021479580.1 | XP_021335255.1 | 47 - 48 | 6349 | 110 | ||
XM_021479581.1 | XP_021335256.1 | 47 - 48 | 6307 | 110 | ||
XM_021479582.1 | XP_021335257.1 | 46 - 47 | 6002 | 110 | ||
XM_021479583.1 | XP_021335258.1 | 47 - 48 | 6266 | 110 | ||
XM_021479584.1 | XP_021335259.1 | 46 - 47 | 6274 | 110 | ||
GenBank mRNA | AY528224.1 | - | 47 - 48 | 6162 | 110 | |
BC163679.1 | - | 46 - 47 | 6002 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |