Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rida | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002576.3 | NP_001002576.2 | 1 - 2 | 140 | 59 | |
GenBank mRNA | BC076217.1 | - | 1 - 2 | 261 | 59 | |
BC107641.1 | - | 1 - 2 | 151 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |