Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epb41l3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003987.1 | NP_001003987.1 | 21 - 22 | 2551 | 88 | |
XM_021470144.1 | XP_021325819.1 | 23 - 24 | 3565 | 88 | ||
XM_021470151.1 | XP_021325826.1 | 23 - 24 | 3214 | 88 | ||
XM_021470154.1 | XP_021325829.1 | 21 - 22 | 2557 | 88 | ||
GenBank mRNA | BC080246.1 | - | 21 - 22 | 2551 | 88 | |
BC158197.1 | - | 21 - 22 | 2548 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |