Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012379.1 | NP_001012379.1 | 4 - 5 | 359 | 143 | |
XM_021479170.1 | XP_021334845.1 | 2 - 3 | 239 | 143 | ||
GenBank mRNA | AB195468.1 | - | 4 - 5 | 359 | 143 | |
BC163126.1 | - | 4 - 5 | 444 | 143 | ||
BC163136.1 | - | 4 - 5 | 441 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |