Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acsl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045128.1 | NP_001038593.1 | 9 - 10 | 1445 | 100 | |
XM_005159236.4 | XP_005159293.1 | 11 - 12 | 1953 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |