Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ensaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045184.1 | NP_001038649.1 | 1 - 2 | 51 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |