Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075103.1 | NP_001068571.1 | 1 - 2 | 706 | 89 | |
GenBank mRNA | BC124299.1 | - | 1 - 2 | 706 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |