Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hapln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082816.2 | NP_001076285.2 | 4 - 5 | 649 | 89 | |
GenBank mRNA | BC163010.1 | - | 4 - 5 | 89 | ||
BC163013.1 | - | 4 - 5 | 89 | |||
CU458972.1 | - | 4 - 5 | 649 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |