Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crabp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182859.1 | NP_878279.1 | 3 - 4 | 645 | 75 | |
GenBank mRNA | AF497478.1 | - | 3 - 4 | 645 | 75 | |
BC091960.1 | - | 3 - 4 | 666 | 75 | ||
BC164791.1 | - | 3 - 4 | 416 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |