Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnot3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199840.1 | NP_956134.1 | 6 - 7 | 551 | 103 | |
XM_009294029.3 | XP_009292304.1 | 6 - 7 | 571 | 103 | ||
XM_017352187.2 | XP_017207676.1 | 6 - 7 | 564 | 103 | ||
GenBank mRNA | AY391415.1 | - | 6 - 7 | 469 | 103 | |
BC045499.1 | - | 6 - 7 | 551 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |