Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002323.1 | NP_001002323.1 | 2 - 3 | 248 | 68 | |
GenBank mRNA | BC076454.1 | - | 2 - 3 | 248 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |