Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044928.1 | NP_001038393.1 | 6 - 7 | 724 | 121 | |
XM_005159517.4 | XP_005159574.1 | 6 - 7 | 755 | 121 | ||
GenBank mRNA | BC171436.1 | - | 6 - 7 | 676 | 121 | |
FN428734.1 | - | 3 - 4 | 249 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |