Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104937.1 | NP_001098407.1 | 14 - 15 | 1730 | 109 | |
XM_005165296.4 | XP_005165353.1 | 14 - 15 | 1798 | 109 | ||
GenBank mRNA | AM084880.1 | - | 14 - 15 | 1730 | 109 | |
AM749801.1 | - | 14 - 15 | 1730 | 109 | ||
AY324390.1 | - | 4 - 5 | 385 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |