Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pax6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131641.1 | NP_571716.1 | 5 - 6 | 670 | 68 | |
XM_005168819.4 | XP_005168876.1 | 6 - 7 | 736 | 68 | ||
XM_005168822.4 | XP_005168879.1 | 6 - 7 | 737 | 68 | ||
XM_005168823.4 | XP_005168880.1 | 3 - 4 | 633 | 68 | ||
XM_009302973.3 | XP_009301248.1 | 5 - 6 | 697 | 68 | ||
XM_017356906.2 | XP_017212395.1 | 5 - 6 | 678 | 68 | ||
XM_021477600.1 | XP_021333275.1 | 6 - 7 | 678 | 68 | ||
XM_021477601.1 | XP_021333276.1 | 7 - 8 | 807 | 68 | ||
XM_021477602.1 | XP_021333277.1 | 6 - 7 | 633 | 68 | ||
XM_021477603.1 | XP_021333278.1 | 6 - 7 | 708 | 68 | ||
GenBank mRNA | AF061252.1 | - | 5 - 6 | 670 | 68 | |
BC076068.1 | - | 5 - 6 | 691 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |