Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201161.1 | NP_957455.1 | 7 - 8 | 653 | 90 | |
XM_021468025.1 | XP_021323700.1 | 8 - 9 | 779 | 90 | ||
XM_021468026.1 | XP_021323701.1 | 7 - 8 | 784 | 90 | ||
XM_021468027.1 | XP_021323702.1 | 8 - 9 | 779 | 90 | ||
XM_021468028.1 | XP_021323703.1 | 7 - 8 | 784 | 90 | ||
GenBank mRNA | BC049418.1 | - | 7 - 8 | 653 | 90 | |
BC097041.1 | - | 7 - 8 | 769 | 90 | ||
KC513820.1 | - | 7 - 8 | 716 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |