Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stk38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213456.1 | NP_998621.1 | 11 - 12 | 1143 | 87 | |
GenBank mRNA | BC044428.1 | - | 11 - 12 | 1143 | 87 | |
BC165742.1 | - | 10 - 11 | 1110 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |