Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syt9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003985.1 | NP_001003985.1 | 6 - 7 | 1696 | 110 | |
GenBank mRNA | BC080256.1 | - | 6 - 7 | 1696 | 110 | |
BC163971.1 | - | 6 - 7 | 1515 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |