Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il12ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007108.1 | NP_001007109.1 | 5 - 6 | 713 | 105 | |
GenBank mRNA | AB183002.1 | - | 5 - 6 | 713 | 105 | |
BC121764.1 | - | 5 - 6 | 712 | 105 | ||
BC164577.1 | - | 4 - 5 | 684 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |