Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacna2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044960.2 | NP_001038425.2 | 11 - 12 | 1130 | 92 | |
XM_005164746.4 | XP_005164803.1 | 11 - 12 | 1300 | 92 | ||
XM_009300285.3 | XP_009298560.1 | 11 - 12 | 1302 | 92 | ||
XM_009300286.3 | XP_009298561.1 | 11 - 12 | 1302 | 92 | ||
XM_009300287.3 | XP_009298562.1 | 11 - 12 | 1300 | 92 | ||
XM_009300288.3 | XP_009298563.1 | 11 - 12 | 1302 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |