Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130932.2 | NP_571007.2 | 4 - 5 | 586 | 64 | |
GenBank mRNA | AF134851.1 | - | 4 - 5 | 512 | 64 | |
AY036972.1 | - | 4 - 5 | 540 | 64 | ||
BC056706.1 | - | 4 - 5 | 558 | 64 | ||
BC065844.1 | - | 4 - 5 | 558 | 64 | ||
EH439264.1 | - | 4 - 5 | 578 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |