Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: cldn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131770.1 | NP_571845.1 | 1 - 2 | 224 | 98 | |
GenBank mRNA | AF359436.1 | - | 1 - 2 | 224 | 98 | |
BC070021.1 | - | 231 | 98 | |||
BC097096.1 | - | 1 - 2 | 265 | 98 | ||
BC098526.1 | - | 1 - 2 | 247 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |