Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200750.1 | NP_957044.1 | 2 - 3 | 188 | 130 | |
GenBank mRNA | BC059557.1 | - | 2 - 3 | 188 | 130 | |
BC150459.1 | - | 3 - 4 | 205 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |