Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205729.2 | NP_991292.2 | 2 - 3 | 638 | 64 | |
GenBank mRNA | AY391444.1 | - | 2 - 3 | 439 | 64 | |
BC095127.1 | - | 2 - 3 | 607 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |